D-(-)-N-Methylglucamine ≥99.0% for resolution of racemates, for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol

M-Clarity™ quality level = MQ200

8.18318.0250
EM8.18318.0250CS 1859 USD
EM8.18318.0250
D-(-)-N-Methylglucamine ≥99.0% for resolution of racemates, for synthesis, Sigma-Aldrich®
D(-)-N-Methylglucamine
Formula: H₃CNHCH₂(CH(OH))₄CH₂OH
MDL Number: MFCD00004707
CAS Number: 6284-40-8

Order Now

Specification Test Results

Solubility 1000 g/l (25 °C)
Melting Point 128 - 131 °C
Molar Mass 195.21 g/mol
Bulk Density 300 kg/m3
Boiling Point 210 °C (1013 hPa) decomposes
pH 11 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR