Sulfur trioxide pyridine complex for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.18112.0100
EM8.18112.0100CS 1296 USD
EM8.18112.0100
Sulfur trioxide pyridine complex for synthesis, Sigma-Aldrich®
Sulphur trioxide pyridine complex
Formula: C₅H₅NO₃S
MDL Number: MFCD00012437
CAS Number: 26412-87-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 155 - 165 °C
Molar Mass 159.16 g/mol
Bulk Density 600 kg/m3
pH (H2O) acidic
Ignition Point 350 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR