Azidotrimethylsilane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Trimethylsilyl azide

M-Clarity™ quality level = MQ200

8.18026.0025
EM8.18026.0025CS 11300 USD
EM8.18026.0025
Azidotrimethylsilane for synthesis, Sigma-Aldrich®
Azidotrimethylsilane
Formula: C₃H₉N₃Si
MW: 115.21 g/mol
MDL Number: MFCD00001986
CAS Number: 4648-54-8
UN: 1992
ADR: 3 (6.1) II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Molar Mass 115.21 g/mol
Boiling Point 92 - 95 °C
Flash Point 6 °C
Refractive Index 1.4142 (20 °C, 589 nm)
Density 0.88 g/cm3 (20 °C)
Ignition Point 300 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR