Oxalic acid, anhydrous for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.16144.0050
EM8.16144.0050CS 6426 USD
EM8.16144.0050
Oxalic acid, anhydrous for synthesis, Sigma-Aldrich®
Oxalic acid
Formula: C₂H₂O₄
Density: 1.9 g/cm³ (20 °C)
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay(Manganometric) ≥99.0%
Water (K F) ≤2.0%
Identity( IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR