L(+)-Lysine monohydrochloride ≥85% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Lysine Hydrochloride, (S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, H-Lys-OH·HCl, L-Lysine monohydrocholoride, L-Lysine Hydrochloride, (S)-(+)-Lysine monohydrochloride, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride, H-Lys-OH.HCl

M-Clarity™ quality level = MQ200

8.16018.0100 8.16018.0500
EM8.16018.0100CS 4841 USD
EM8.16018.0100 EM8.16018.0500
L(+)-Lysine monohydrochloride ≥85% for synthesis, Sigma-Aldrich®
L(+)-Lysine monohydrochloride
Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MW: 182.65 g/mol
MDL Number: MFCD00064564
CAS Number: 657-27-2

Order Now

Specification Test Results

Solubility 420 g/l (20 °C)
Melting Point 263 - 264 °C
Molar Mass 182.64 g/mol
Bulk Density 360 kg/m3
pH 5.5 - 6.0 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR