Glycine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

M-Clarity™ quality level = MQ200

8.16013.0250 8.16013.1000
EM8.16013.0250CS 1135 USD
EM8.16013.0250 EM8.16013.1000
Glycine for synthesis, Sigma-Aldrich®
Glycine
Formula: NH₂CH₂COOH
MDL Number: MFCD00008131
CAS Number: 56-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 225 g/l (20 °C)
Melting Point 232 - 236 °C (decomposition)
Molar Mass 75.06 g/mol
Bulk Density 920 kg/m3
Density 1.595 g/cm3 (20 °C)
pH 5.9 - 6.4 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR