L-Phenylalanine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.16011.0010 8.16011.0050
EM8.16011.0010CS 3223 USD
EM8.16011.0010 EM8.16011.0050
L-Phenylalanine for synthesis, Sigma-Aldrich®
L-α-Phenylalanine
Formula: C₉H₁₁NO₂
MDL Number: MFCD00064227
CAS Number: 63-91-2

Order Now

Specification Test Results

Solubility 27 g/l (20 °C)
Melting Point 275 - 283 °C (decomposition)
Molar Mass 165.19 g/mol
Bulk Density 580 kg/m3
pH 5.4 - 6.0 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR