L(+)-Glutamic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid

M-Clarity™ quality level = MQ200

8.16004.1000
EM8.16004.1000CS 1009 USD
EM8.16004.1000
L(+)-Glutamic acid for synthesis, Sigma-Aldrich®
L(+)-Glutamic acid
Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
MW: 147.13 g/mol
MDL Number: MFCD00002634
CAS Number: 56-86-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (acidimetric) ≥ 99.0 %
Identity (IR) passes test
Spec. rotation α²0/D (c=10 in HCl 2 mol/l) +30.0 - +32.5 °

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR