(S)-(+)-Arginine hydrochloride ≥99% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: H-L-Arg-OH.HCl, (S)-2-Amino-5-guanidinopentanoic acid hydrochloride, (S)-(+)-Arginine HCl, H-Arg-OH·HCl, L-Arginine, hydrochloride, (S)-(+)-Arginine hydrochloride, H-Arg-OH.HCl, L-Arg, L(+)-Arginine monohydrochloride, L(+)-Arginine hydrochloride

M-Clarity™ quality level = MQ200

8.16001.0100
EM8.16001.0100CS 3688 USD
EM8.16001.0100
(S)-(+)-Arginine hydrochloride ≥99% for synthesis, Sigma-Aldrich®
L-Arginine hydrochloride
Formula: C₆H₁₅ClN₄O₂
MW: 210.66 g/mol
MDL Number: MFCD00064550
CAS Number: 1119-34-2

Order Now

Specification Test Results

Assay (argentometric) ≥ 99.0 %
Spec. rotation [α²0/D (c=8 in HCl 6 mol/l) +21.5 - +23.0 °
Identity (IR) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR