4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222), special quality for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Cryptand 2.2.2, Fludeoxyglucose Related Compound A, Kryptofix 222, 4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo(8.8.8)hexacosane, 4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane, Cryptand 222, (2.2.2)Cryptand

M-Clarity™ quality level = MQ200

8.14925.0001
EM8.14925.0001CS 53727 USD
EM8.14925.0001
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222), special quality for synthesis, Sigma-Aldrich®
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222)
Formula: C₁₈H₃₆N₂O₆
MW: 376.49 g/mol
MDL Number: MFCD00005111
CAS Number: 23978-09-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a74ac532561e4fecf111f3260b402bcd1fb0af9&vid=13491901da172b5971e6d7f330c893460a77cd23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Melting Point 68 - 71 °C
Molar Mass 376.49 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR