Tetrakis(triphenylphosphine)palladium(0), Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4

M-Clarity™ quality level = MQ200

8.14761.8500
EM8.14761.8500EA 113 USD
EM8.14761.8500
Tetrakis(triphenylphosphine)palladium(0), Sigma-Aldrich®
Tetrakis(triphenylphosphine)palladium (0)
Formula: C₇₂H₆₀P₄Pd
Melting Pt: 103…107 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00010012
CAS Number: 14221-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR