DL-Malic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.14737.1000 8.14737.5000
EM8.14737.1000CS 541 USD
EM8.14737.1000 EM8.14737.5000
DL-Malic acid for synthesis, Sigma-Aldrich®
DL-Malic acid
Formula: HOOCCH(OH)CH₂COOH
MDL Number: MFCD00064212
CAS Number: 617-48-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=920b54632a869bc5df22624fb9fecd4d11cae65e&vid=278193678126274041defdc0bd71c7c40efb3085&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (acidimetric) ≥ 99.0 %
Identity (IR) passes test
Melting range (lower value) ≥ 128 °C
Melting range (upper value) ≤ 132 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR