4-Amino-2,2,6,6-tetramethylpiperidine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.14722.0100
EM8.14722.0100CS 1532 USD
EM8.14722.0100
4-Amino-2,2,6,6-tetramethylpiperidine for synthesis, Sigma-Aldrich®
4-Amino-2,2,6,6-tetramethylpiperidine
Formula: C₉H₂₀N₂
MDL Number: MFCD00005984
CAS Number: 36768-62-4
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 12 - 15 °C
Molar Mass 156.27 g/mol
Boiling Point 199 °C (1013 hPa)
Vapor Pressure = - 20 hPa (20 °C)
Flash Point 75 °C
Refractive Index 1.469 (25 °C)
Density 0.893 g/cm3 (20 °C)
pH 12.3 (99 g/l, H2O, 20 °C)
Ignition Point 300 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR