Copper(II) bromide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Cupric bromide, Cuprous bromide

M-Clarity™ quality level = MQ200

8.14658.0050 8.14658.0250
EM8.14658.0050CS 1789 USD
EM8.14658.0050 EM8.14658.0250
Copper(II) bromide for synthesis, Sigma-Aldrich®
Copper (II) bromide
Formula: Br₂Cu
MW: 223.35 g/mol
Boiling Pt: 900
Melting Pt: 498
MDL Number: MFCD00010970
CAS Number: 7789-45-9
UN: 1759
ADR: 8 II
Merck Index: 13,02656

Order Now

Specification Test Results

Solubility 1200 g/l (25 °C)
Melting Point 498 °C
Molar Mass 223.35 g/mol
Bulk Density 2000 kg/m3
Boiling Point 900 °C (1013 hPa)
Density 4.71 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR