Sodium peroxide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: di-Sodium peroxide

M-Clarity™ quality level = MQ200

8.14606.0100
EM8.14606.0100CS 8715 USD
EM8.14606.0100
Sodium peroxide for synthesis, Sigma-Aldrich®
Sodium peroxide
Formula: Na₂O₂
MW: 77.98 g/mol
MDL Number: MFCD00003497
CAS Number: 1313-60-6
UN: 1504
ADR: 5.1,I

Order Now

Specification Test Results

Solubility 100 g/l (20 °C) (decomposition)
Melting Point 660 °C
Molar Mass 77.98 g/mol
Bulk Density 1138 kg/m3
Density 2.8 g/cm3 (20 °C)
pH 12.8 (100 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR