Amidosulfonic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Aminosulphonic acid, Amidosulphuric acid, Sulfamic acid, Sulfamidic acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid, Aminosulfonic acid, Amidosulfonic acid, Amidosulfuric Acid, Amidosulfuric acid
M-Clarity™ quality level = MQ200
8.14483.0100
8.14483.1000
EM8.14483.0100CS
3325
USD
EM8.14483.0100
EM8.14483.1000
Amidosulfonic acid for synthesis, Sigma-Aldrich®
Amidosulphonic acid
Formula:
H₃NO₃S MW: 97.09 g/mol Boiling Pt: 247 Melting Pt: 205 |
MDL Number:
MFCD00011603 CAS Number: 5329-14-6 UN: 2967 ADR: 8 III Merck Index: 13,09006 |
|
Specification Test Results
Assay (acidimetric) | ≥ 99.0 % |
Identity (IR) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...