Triethylammonium dihydrogentrifluoride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Triethylamine trihydrofluoride, Triethylamine trishydrofluoride

M-Clarity™ quality level = MQ200

8.14371.0010
EM8.14371.0010CS 35557 USD
EM8.14371.0010
Triethylammonium dihydrogentrifluoride for synthesis, Sigma-Aldrich®
Triethylammonium dihydrogentrifluoride
Formula: C₆H₁₈F₃N
MW: 161.21
Density: 1.094
MDL Number: MFCD00043294
CAS Number: 73602-61-6
UN: 2927
ADR: 6.1 (8) I

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 161.21 g/mol
Boiling Point 70 °C (19.9 hPa)
Flash Point 88 °C
Density 1.00 g/cm3 (20 °C)
pH 3 (H2O, 20 °C) (saturated aqueous solution)
Hill Formula C6H15N*3FH
Chemical Formula (C2H5)3N * 3 HF


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR