Triethylammonium dihydrogentrifluoride, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Triethylamine trihydrofluoride, Triethylamine trishydrofluoride

M-Clarity™ quality level = MQ200

8.14371.0002
EM8.14371.0002EA 29 USD
EM8.14371.0002
Triethylammonium dihydrogentrifluoride, Sigma-Aldrich®
Triethylammonium dihydrogentrifluoride
Formula: C₆H₁₈F₃N
MW: 161.21
Density: 1.094 g/cm³ (20 °C)
MDL Number: MFCD00043294
CAS Number: 73602-61-6
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR