Lithium tetrafluoroborate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.14347.0010
EM8.14347.0010CS 54230 USD
EM8.14347.0010
Lithium tetrafluoroborate for synthesis, Sigma-Aldrich®
Lithium tetrafluoroborate
Formula: BF₄Li
MW: 93.74 g/mol
MDL Number: MFCD00011087
CAS Number: 14283-07-9
UN: 3260
ADR: 8 II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 324 °C (decomposition)
Molar Mass 93.74 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR