Pyridinium tribromide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Pyridinium bromide perbromide, Monopyridin-1-ium tribromide, Pyridine hydrobromide perbromide

M-Clarity™ quality level = MQ200

8.14345.0050
EM8.14345.0050CS 10984 USD
EM8.14345.0050
Pyridinium tribromide for synthesis, Sigma-Aldrich®
Pyridinium tribromide
Formula: C₅H₆Br₃N
MW: 319.82 g/mol
MDL Number: MFCD00013223
CAS Number: 39416-48-3
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Solubility 0.17 g/l (20 °C)
Melting Point -8 °C
Molar Mass 183.12 g/mol
Boiling Point 240 - 242 °C (1013 hPa)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 118 °C
Refractive Index 1.457 (25 °C, 589 nm)
Density 1.03 g/cm3 (20 °C)
Ignition Point 215 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR