Bromotrimethylsilane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Trimethylbromosilane, Trimethylsilyl bromide

M-Clarity™ quality level = MQ200

8.14324.0010 8.14324.0100
EM8.14324.0010CS 77445 USD
EM8.14324.0010 EM8.14324.0100
Bromotrimethylsilane for synthesis, Sigma-Aldrich®
Bromotrimethylsilane
Formula: C₃H₉BrSi
MW: 153.09 g/mol
Boiling Pt: 79
Melting Pt: -43
MDL Number: MFCD00000048
CAS Number: 2857-97-8
UN: 2920
ADR: 8 (3) II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point -43 °C
Molar Mass 153.09 g/mol
Boiling Point 79 °C
Flash Point 32 °C
Refractive Index 1.4211 (25 °C, 589 nm)
Density 1.17 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR