Iminodiacetic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2,2'-Azanediyldiacetic acid

M-Clarity™ quality level = MQ200

8.14165.0250
EM8.14165.0250CS 1854 USD
EM8.14165.0250
Iminodiacetic acid for synthesis, Sigma-Aldrich®
Iminodiacetic acid
Formula: C₄H₇NO₄
MDL Number: MFCD00004280
CAS Number: 142-73-4

Order Now

Specification Test Results

Solubility 42 g/l (20 °C)
Melting Point 247 °C (decomposition)
Molar Mass 133.10 g/mol
pH 2.2 - 2.3 (H2O, 20 °C) (saturated solution)
Hill Formula C4H7NO4
Chemical Formula HN(CH2COOH)2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR