2,5-Piperazinedione for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.14160.0025
EM8.14160.0025CS 11300 USD
EM8.14160.0025
2,5-Piperazinedione for synthesis, Sigma-Aldrich®
2,5-Piperazinedione
Formula: C₄H₆N₂O₂
MDL Number: MFCD00006009
CAS Number: 106-57-0

Order Now

Specification Test Results

Solubility 142 g/l (20 °C)
Molar Mass 114.1 g/mol
Synonyms Glycinanhydride, 2,5-Diketopiperazine
Hill Formula C4H6N2O2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR