Cetrimonium chloride 50% in 2-propanol/water (2:3) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Cetyltrimethylammonium chloride, (1-Hexadecyl)trimethylammonium chloride, Hexadecyltrimethylammonium chloride, N-Cetyl-N,N,N-trimethylammonium chloride, N,N,N-Trimethylhexadecan-1-aminium chloride, CTAC
M-Clarity™ quality level = MQ200
8.14087.0100
8.14087.1000
EM8.14087.0100CS
621
USD
EM8.14087.0100
EM8.14087.1000
Cetrimonium chloride 50% in 2-propanol/water (2:3) for synthesis, Sigma-Aldrich®
Cetrimonium chloride
Formula:
C₁₉H₄₂ClN |
MDL Number:
MFCD00011773 CAS Number: 112-02-7 UN: 2924 ADR: 3 (8),II |
Specification Test Results
Solubility | (20 °C) soluble |
Boiling Point | 80 - 90 °C |
Flash Point | 15 °C |
Refractive Index | 1.3778 (20 °C, 589 nm) |
Density | 0.89 g/cm3 (20 °C) |
pH | 5 - 7 (10 g/l, H2O, 20 °C) |
Synonyms | Cetyltrimethylammonium chloride solution, Trimethylhexadecylammonium chloride |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...