Ruthenium(III) acetylacetonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Ruthenium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')ruthenium (III), Ruthenium (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato)ruthenium(III)

M-Clarity™ quality level = MQ200

8.14056.0001 8.14056.0005
EM8.14056.0001CS 186423 USD
EM8.14056.0001 EM8.14056.0005
Ruthenium(III) acetylacetonate for synthesis, Sigma-Aldrich®
Ruthenium (III) acetylacetonate
Formula: C₁₅H₂₁O₆Ru
MDL Number: MFCD00000030
CAS Number: 14284-93-6

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 230 - 235 °C
Molar Mass 398.39 g/mol
Synonyms Tris(acetylacetonato)ruthenium(III), Tris(pentane-2,4-dionato)ruthenium(III)
Hill Formula C15H21O6Ru
Chemical Formula (CH3C(O)=CHCOCH3)3Ru


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR