Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Dibenzo-18-crown-6

M-Clarity™ quality level = MQ200

8.11731.0005 8.11731.0050
EM8.11731.0005CS 2801 USD
EM8.11731.0005 EM8.11731.0050
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) for synthesis, Sigma-Aldrich®
Dibenzo-18-crown-6 (Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin (II) ionophore I)
Formula: C₂₀H₂₄O₆
MDL Number: MFCD00005098
CAS Number: 14187-32-7

Order Now

Specification Test Results

Solubility 0.007 g/l (25 °C)
Melting Point 158 - 161 °C
Molar Mass 360.41 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR