8,8'-[Oxybis(ethyleneoxyethyleneoxy)]diquinoline (Kryptofix® 5) ≥99.0% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 1,13-Bis(8-quinolyl)-1,4,7,10,13-pentaoxatridecane

M-Clarity™ quality level = MQ200

8.11689.0005
EM8.11689.0005CS 10452 USD
EM8.11689.0005
8,8'-[Oxybis(ethyleneoxyethyleneoxy)]diquinoline (Kryptofix® 5) ≥99.0% for synthesis, Sigma-Aldrich®
8,8'-[Oxybis(ethyleneoxyethyleneoxy)]diquinoline (Kryptofix® 5)
Formula: C₂₆H₂₈N₂O₅
MW: 448.52 g/mol
MDL Number: MFCD00040469
CAS Number: 57310-75-5

Order Now

Specification Test Results

Assay (HClO₄) ≥ 99.0 %
Melting range (lower value) ≥ 72 °C
Melting range (upper value) ≤ 76 °C
Identity (IR) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR