Thionyl chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.08154.0500 8.08154.1000 8.08154.2500
EM8.08154.0500CS 2602 USD
EM8.08154.0500 EM8.08154.1000 EM8.08154.2500 EM8.08154.9040
Thionyl chloride for synthesis, Sigma-Aldrich®
Thionyl chloride
Formula: Cl₂OS
Boiling Pt: 76 °C (1013 hPa)
Melting Pt: –104.5 °C
Density: 1.64 g/cm³ (20 °C)
MDL Number: MFCD00011449
CAS Number: 7719-09-7
EINECS: 231-748-8
UN: 1836
ADR: 8,I
Merck Index: 12,09485

Order Now

Specification Test Results

Solubility (20 °C) (rigorous decomposition)
Melting Point -104.5 °C
Molar Mass 118.97 g/mol
Boiling Point 76 °C (1013 hPa)
Vapor Pressure 121 hPa (20 °C)
Refractive Index 1.517 (20 °C, 589 nm)
Density 1.64 g/cm3 (20 °C)
pH (H2O) not applicable
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR