Sulfolane ≥99.0% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.07993.0250
EM8.07993.0250CS 912 USD
EM8.07993.0250
Sulfolane ≥99.0% for synthesis, Sigma-Aldrich®
Sulpholane
Formula: C₄H₈O₂S
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 26 °C
Density: 1.26 g/cm3 (30 °C)
Flash Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00005484
CAS Number: 126-33-0
EINECS: 204-783-1
ADR: 9,III

Order Now

Specification Test Results

Assay (GC, area%) ≥ 99.0 % (a/a)
Water (K. F.) ≤ 0.20 %
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR