Pyrrolidine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.07494.0005
8.07494.1000
EM8.07494.0005CS
2933
USD
EM8.07494.0005
EM8.07494.1000
Pyrrolidine for synthesis, Sigma-Aldrich®
Pyrrolidine
Formula:
C₄H₉N Boiling Pt: 87 °C (1013 hPa) Melting Pt: –63 °C Density: 0.86 g/cm³ (20 °C) Flash Pt: 3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005249 CAS Number: 123-75-1 EINECS: 204-648-7 UN: 1922 ADR: 3 (8),II Merck Index: 12,08199 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -63 °C |
Molar Mass | 71.12 g/mol |
Boiling Point | 86 - 88 °C (1013 hPa) |
Vapor Pressure | 65 hPa (20 °C) |
Flash Point | 3 °C |
Refractive Index | 1.4435 (20 °C, 589 nm) |
Explosion Limit | 1.6 - 10.6 %(V) |
Density | 0.86 g/cm3 (20 °C) |
pH | 12.9 (100 g/l, H2O, 20 °C) |
Ignition Point | 345 °C |
Saturation concentration | 466 g/m3 (20 °C) Air |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...