Polyethylene glycol 1,000 for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: PEG 1,000, PEG 900-1100

M-Clarity™ quality level = MQ200

8.07488.1000 8.07488.5000
EM8.07488.1000CS 457 USD
EM8.07488.1000 EM8.07488.5000
Polyethylene glycol 1,000 for synthesis, Sigma-Aldrich®
Polyethylene glycol 1,000
Formula: H(OCH₂CH₂)nOH
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 33…40 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 13,07651

Order Now

Specification Test Results

Solubility 750 g/l (20 °C)
Melting Point 33 - 40 °C
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 260 °C
Density 1.2 g/cm3 (20 °C)
pH 4 - 7 (100 g/l, H2O, 20 °C)
Ignition Point 420 °C
Viscosity kinematic 22 - 27 mm2/s (20 °C) (50% solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR