Polyethylene glycol 400 for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: PEG 400

M-Clarity™ quality level = MQ200

8.07485.5000
EM8.07485.5000CS 629 USD
EM8.07485.5000
Polyethylene glycol 400 for synthesis, Sigma-Aldrich®
Polyethylene glycol 400
Formula: H(OCH₂CH₂)nOH
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 4…8 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 245 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 4 - 8 °C
Molar Mass 8000 g/mol
Boiling Point 200 °C (1013 hPa)
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 240 °C
Density 1.13 g/cm3 (20 °C)
pH 5 - 7 (100 g/l, H2O, 20 °C)
Ignition Point 360 °C DIN 51794
Viscosity kinematic 97 - 110 mm2/s (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR