Polyphosphoric acid, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.07471.9025
EM8.07471.9025EA 2296 USD
EM8.07471.9025
Polyphosphoric acid, Sigma-Aldrich®
Polyphosphoric acid
Formula: H(PO₃H)nOH
MDL Number: MFCD00084480
CAS Number: 8017-16-1
UN: 3264
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR