Polyphosphoric acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.07471.0100 8.07471.0500 8.07471.2500
EM8.07471.0100CS 9731 USD
EM8.07471.0100 EM8.07471.0500 EM8.07471.2500
Polyphosphoric acid for synthesis, Sigma-Aldrich®
Polyphosphoric acid
Formula: H(PO₃H)nOH
MDL Number: MFCD00084480
CAS Number: 8017-16-1
UN: 3264
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble,Hydrolysis
Melting Point -20 °C
Boiling Point 530 °C (1013 hPa)
Vapor Pressure 2 hPa (20 °C)
Density 2.06 g/cm3 (20 °C)
pH (H2O, 20 °C) acidic,Hydrolysis
Synonyms PPA
Hill Formula HO[P(OH)(O)O](n)H


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR