Hexachloroplatinic(IV) acid hexahydrate Pt ∼ 40% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Hydrogen hexachloroplatinate (IV) hexahydrate, Chloroplatinic acid hexahydrate, Dihydrogen hexachloroplatinate (IV) hexahydrate

M-Clarity™ quality level = MQ200

8.07340.0001 8.07340.0005 8.07340.0025 8.07340.8500
EM8.07340.0001CS 32638 USD
EM8.07340.0001 EM8.07340.0005 EM8.07340.0025 EM8.07340.8500
Hexachloroplatinic(IV) acid hexahydrate Pt ∼ 40% for synthesis, Sigma-Aldrich®
Hexachloroplatinic (IV) acid hexahydrate
Formula: H₂PtCl₆·6H₂O
Melting Pt: 150 °C
Density: 2.434 g/cm³ (20 °C)
MDL Number: MFCD00149910
CAS Number: 18497-13-7
EINECS: 241-010-7
UN: 2507
ADR: 8,III
Merck Index: 13,07609

Order Now

Specification Test Results

Melting Point 150 °C
Molar Mass 517.94 g/mol
Density 4 g/cm3 (20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR