Platinum 10% on activated carbon hydrogenation catalyst, for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.07339.0001 8.07339.0005
EM8.07339.0001CS 48974 USD
EM8.07339.0001 EM8.07339.0005
Platinum 10% on activated carbon hydrogenation catalyst, for synthesis, Sigma-Aldrich®
Platinum
Formula: Pt
MDL Number: MFCD00011179
CAS Number: 7440-06-4
UN: 3088
ADR: 4.2,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR