Pentaerythritol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.07331.0100 8.07331.1000
EM8.07331.0100CS 6206 USD
EM8.07331.0100 EM8.07331.1000
Pentaerythritol for synthesis, Sigma-Aldrich®
Pentaerythritol
Formula: C₅H₁₂O₄
MW: 136.15 g/mol
Boiling Pt: 380
Melting Pt: 260…264 °C
Density: 1.511 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004692
CAS Number: 115-77-5
EINECS: 204-104-9
Merck Index: 12,07245

Order Now

Specification Test Results

Solubility 72.3 g/l (25 °C)
Melting Point 256 - 258 °C
Molar Mass 136.14 g/mol
Bulk Density 750 - 850 kg/m3
Boiling Point 276 °C (40 hPa)
Vapor Pressure  - 0.00001 hPa (20 °C)
Density 1.39 g/cm3 (20 °C)
pH 3.5 - 4.5 (100 g/l, H2O, 35 °C)
Ignition Point 490 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR