Pyrazine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.07064.0100
EM8.07064.0100CS 3371 USD
EM8.07064.0100
Pyrazine for synthesis, Sigma-Aldrich®
Pyrazine
Formula: C₄H₄N₂
MDL Number: MFCD00006122
CAS Number: 290-37-9
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 49 - 52 °C
Molar Mass 80.08 g/mol
Boiling Point 115 - 118 °C (1013 hPa)
Synonyms 1,4-Diazine
Hill Formula C4H4N2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR