Ethyl nicotinate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.06754.0250
EM8.06754.0050CS 3489.64 USD
EM8.06754.0050 EM8.06754.0250
Ethyl nicotinate for synthesis, Sigma-Aldrich®
Ethyl nicotinate
Formula: C₈H₉NO₂
MDL Number: MFCD00006389
CAS Number: 614-18-6

Order Now

Specification Test Results

Solubility 50 g/l (20 °C)
Melting Point 8 - 9 °C
Molar Mass 151.16 g/mol
Boiling Point 103 - 105 °C (7 hPa)
Flash Point 93 °C
Refractive Index 1.504 (20 °C, 589 nm)
Density 1.11 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR