Methylcyclohexane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.06147.0100 8.06147.1000 8.06147.2500
EM8.06147.0100CS 747 USD
EM8.06147.0100 EM8.06147.1000 EM8.06147.2500
Methylcyclohexane for synthesis, Sigma-Aldrich®
Methylcyclohexane
Formula: C₆H₁₁CH₃
MDL Number: MFCD00001497
CAS Number: 108-87-2
UN: 2296
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f7f0d5a1c32138da16df4781897a9c0184e42b7&vid=39a9392c8629d0e2fc79134840e6d8a8dc8a76e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.014 g/l (20 °C)
Melting Point -126 °C
Molar Mass 98.19 g/mol
Boiling Point 100.9 °C (1013 hPa)
Vapor Pressure 48 hPa (20 °C)
Flash Point -4 °C
Explosion Limit 1.1 - 6.7 %(V)
Density 0.77 g/cm3 (20 °C)
Ignition Point 260 °C
Hill Formula C7H14
Chemical Formula C6H11CH3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR