N-Methyl-2-pyrrolidone (NMP), EMPLURA®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone
8.06072.1000
8.06072.2500
8.06072.9011
8.06072.9025
EM8.06072.1000CS
1458
USD
EM8.06072.1000
EM8.06072.2500
EM8.06072.9011
EM8.06072.9025
N-Methyl-2-pyrrolidone (NMP), EMPLURA®, Supelco®
N-Methyl-2-pyrrolidone (NMP)
Formula:
C₅H₉NO MW: 99.13 g/mol Boiling Pt: 202 Melting Pt: -24 Density: 1.033 g/cm³ (20 °C) Flash Pt: 91 Storage Temperature: Ambient |
MDL Number:
MFCD00003193 CAS Number: 872-50-4 EINECS: 212-828-1 Merck Index: 13,06140 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...