Bisacrylamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS

M-Clarity™ quality level = MQ200

8.05968.0250
EM8.05968.0250CS 1214 USD
EM8.05968.0250
Bisacrylamide for synthesis, Sigma-Aldrich®
Bisacrylamide
Formula: (H₂C=CHCONH)₂CH₂
MW: 154.17 g/mol
Boiling Pt: 125 °C (3.33 hPa)
Melting Pt: 185 °C
Density: 1.235 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008625
CAS Number: 110-26-9
EINECS: 203-750-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b72b0c3cd5bb916c65f9e2337c5c3b05ea1c91&vid=368f6bf0bc8e0b35ffc7a57d85f5dd210ff0b409&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 185 °C (decomposition)
Molar Mass 154.17 g/mol
Bulk Density 200 kg/m3
Density 1.235 g/cm3 (30 °C)
pH =5.0 (25 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR