2-Pentanone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Methyl Propyl Ketone

M-Clarity™ quality level = MQ200

8.05965.0100 8.05965.0500
EM8.05965.0100CS 868 USD
EM8.05965.0100 EM8.05965.0500
2-Pentanone for synthesis, Sigma-Aldrich®
2-Pentanone
Formula: C₅H₁₀O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: –78 °C
Density: 0.809 g/cm³ (20 °C)
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00009400
CAS Number: 107-87-9
EINECS: 203-528-1
UN: 1249
ADR: 3,II
Merck Index: 12,06193

Order Now

Specification Test Results

Solubility 43 g/l (25 °C)
Melting Point -78 °C
Molar Mass 86.13 g/mol
Boiling Point 101 - 103 °C (1013 hPa)
Vapor Pressure 37 hPa (20 °C)
Flash Point 7 °C
Refractive Index 1.3895 (20 °C, 589 nm)
Explosion Limit 1.5 - 8.2 %(V)
Density 0.81 g/cm3 (20 °C)
Ignition Point 449 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR