Manganese(IV) oxide for synthesis, (precipitated active), Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid

M-Clarity™ quality level = MQ200

8.05958.1000
EM8.05958.1000EA 117 USD
EM8.05958.1000
Manganese(IV) oxide for synthesis, (precipitated active), Sigma-Aldrich®
Manganese (IV) oxide
Formula: MnO₂
MW: 86.94 g/mol
Melting Pt: 535 °C
Density: 5.03 g/cm³ (20 °C)
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,05730

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR