α-Ionone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: (E)-4-(2,6,6-Trimethylcyclohex-2-en-1-yl)but-3-en-2-one

M-Clarity™ quality level = MQ200

8.04778.0050
EM8.04778.0050CS 3867 USD
EM8.04778.0050
α-Ionone for synthesis, Sigma-Aldrich®
α-Ionone
Formula: C₁₃H₂₀O
MW: 192.3 g/mol
MDL Number: MFCD00001565
CAS Number: 127-41-3

Order Now

Specification Test Results

Solubility 110 mg/l (20 °C)
Molar Mass 192.3 g/mol
Boiling Point 237 °C (1013 hPa)
Vapor Pressure 0.0013 hPa (20 °C)
Flash Point 117 °C
Refractive Index 1.5 (20 °C, 589 nm)
Density 0.93 g/cm3 (20 °C)
Hill Formula C13H20O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR