Iodoacetamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.04744.0100 8.04744.1000 8.04744.2500
EM8.04744.0100CS 3736 USD
EM8.04744.0100 EM8.04744.1000 EM8.04744.2500
Iodoacetamide for synthesis, Sigma-Aldrich®
Iodoacetamide
Formula: ICH₂CONH₂
MDL Number: MFCD00008028
CAS Number: 144-48-9
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 91 - 94 °C
Molar Mass 184.96 g/mol
Hill Formula C2H4INO
Chemical Formula ICH2CONH2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR