Indole-3-carbaldehyd for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 1H-Indole-3-carbaldehyde, Indole-3-carboxaldehyde

M-Clarity™ quality level = MQ200

8.04726.0025
EM8.04726.0025CS 20728 USD
EM8.04726.0025
Indole-3-carbaldehyd for synthesis, Sigma-Aldrich®
Indole-3-carbaldehyd
Formula: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD00005622
CAS Number: 487-89-8

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 192 - 195 °C
Molar Mass 145.16 g/mol
Bulk Density 200 - 300 kg/m3
Flash Point 240 °C
Ignition Point 550 °C
Synonyms Indol-3-aldehyde
Hill Formula C9H7NO

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR