Dibutylamine, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Di-n-butylamine, N-butyl-1-propanamine

M-Clarity™ quality level = MQ200

8.03222.1000
EM8.03222.1000EA 131 USD
EM8.03222.1000
Dibutylamine, Sigma-Aldrich®
Dibutylamine
Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.759 g/cm³ (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00009429
CAS Number: 111-92-2
EINECS: 203-921-8
UN: 2248
ADR: 8 (3),II
Merck Index: 13,03058
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0d7d86589224f8565e12dc8e6b30a8de923393f&vid=d2cc023db25785e54555fc105aa8436e65fd9cbc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR