Diethanolamine ≥99% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.03116.9022 8.03116.9051
EM8.03116.1000EA 84 USD
EM8.03116.1000 EM8.03116.5000 EM8.03116.9022 EM8.03116.9051
Diethanolamine ≥99% (by GC) for synthesis, Sigma-Aldrich®
Diethanolamine
Formula: C₄H₁₁NO₂
Boiling Pt: 269…270 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.09 g/cm3 at 30 °C (liquid)
Storage Temperature: Ambient
MDL Number: MFCD00002843
CAS Number: 111-42-2
UN: 3077

Order Now

Specification Test Results

Solubility (20°C) Soluble
Melting Point 28 °C
Boiling Point 269 - 270 °C (1013 hPa)
Vapor Pressure  - 0.01 hPa (25 °C)
Flash Point 176 °C
Refractive Index 1.4753 (30 °C, 589 nm)
Explosion Limit 2.1 - 10.6 %(V)
Density 1.09 g/cm3 (30 °C) liquid
pH 11 (53 g/l, H2O, 20 °C)
Ignition Point 365 - 370°C
Synonyms ß,ß-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine
Hill Formula C4H11NO2
Chemical Formula NH(CH2CH2OH)2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR