Cyanoguanidine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.02491.9050
EM8.02491.9050EA 3011 USD
EM8.02491.9050
Cyanoguanidine for synthesis, Sigma-Aldrich®
Cyanoguanidine
Formula: (H₂N)₂CNCN
Melting Pt: 208…210 °C
Density: 1.4 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008066
CAS Number: 461-58-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility 34.6 g/l (20°C)
Melting Point 208 - 210 °C
Molar Mass 400 kg/m3
Bulk Density 400 kg/m3
Density 1.4 g/cm3 (20 °C)
Synonyms Dicyanodiamide
Hill Formula C2H4N4
Chemical Formula NCNHC(=NH)NH2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR